This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05531949_g1
See other CERCAM GE Assays ›
Gene Symbol:
CERCAM
Gene Name:
cerebral endothelial cell adhesion molecule
Gene Aliases:
CEECAM1, GLT25D3
Chromosome Location:
Chr.9: 128405993 - 128437351 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05531949_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CERCAM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017014794.1 XP_016870283.1 2 - 3 223 60

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017014794.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

Gene Ontology Categories:

Function(s)

movement of cell or subcellular component
cell adhesion
leukocyte cell-cell adhesion

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline