This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05549677_g1
See other EIF4B GE Assays ›
Gene Symbol:
EIF4B
Gene Name:
eukaryotic translation initiation factor 4B
Gene Aliases:
EIF-4B, PRO1843
Chromosome Location:
Chr.12: 53006258 - 53042209 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05549677_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF4B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300821.1 NP_001287750.1 15 2727 99
NM_001417.5 NP_001408.2 15 2712 99
GenBank mRNA AF119854.1 - 1 99
AK312187.1 - 1 99
BC001097.1 - 8 1702 99
BC047302.1 - 1 318 99
BC073139.1 - 15 2501 6527540 99
BU902688.1 - 1 279 99
CR627377.1 - 9 1572 99

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EIF4B

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA poly(A) tail shortening
formation of translation preinitiation complex
cytoplasmic translation
translational initiation
regulation of translational initiation
eukaryotic translation initiation factor 4F complex assembly
nucleotide binding
RNA binding
translation initiation factor activity
helicase activity
protein binding
RNA strand annealing activity
RNA strand-exchange activity
ribosomal small subunit binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z47m5:80/100.66.77.18:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0