This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05557243_g1
See other DDX11 GE Assays ›
Gene Symbol:
DDX11
Gene Name:
DEAD/H-box helicase 11
Gene Aliases:
CHL1, CHLR1, KRG2, WABS
Chromosome Location:
Chr.12: 31073574 - 31104799 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05557243_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DDX11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017018937.1 XP_016874426.1 1 151 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017018937.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

nucleobase-containing compound metabolic process
sister chromatid cohesion
viral process
DNA duplex unwinding
IRE1-mediated unfolded protein response
double-stranded DNA binding
single-stranded DNA binding
RNA binding
ATP-dependent DNA helicase activity
helicase activity
protein binding
ATP binding
DNA-dependent ATPase activity
metal ion binding
4 iron, 4 sulfur cluster binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline