This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05563926_s1
See other ETFBKMT GE Assays ›
Gene Symbol:
ETFBKMT
Gene Name:
electron transfer flavoprotein beta subunit lysine methyltransferase
Gene Aliases:
C12orf72, ETFB-KMT, METTL20
Chromosome Location:
Chr.12: 31647160 - 31669082 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05563926_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ETFBKMT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001135863.1 NP_001129335.1 2 210 99
NM_001135864.1 NP_001129336.1 2 191 99
NM_173802.3 NP_776163.1 2 271 99
GenBank mRNA AK290248.1 - 2 192 99
AL832339.1 - 2 197 99
BC039107.1 - 3 4837913 99
BC039535.1 - 2 271 5744381 99
DA303137.1 - 2 210 99

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK290248.1, BC039107.1, BC039535.1, NM_001135863.1, NM_001135864.1, NM_173802.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ETFBKMT

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

peptidyl-lysine methylation
peptidyl-lysine trimethylation
negative regulation of electron carrier activity
negative regulation of fatty acid beta-oxidation using acyl-CoA dehydrogenase
protein-lysine N-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0