This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05568456_s1
See other SDSL GE Assays ›
Gene Symbol:
SDSL
Gene Name:
serine dehydratase like
Gene Aliases:
SDH 2, SDS-RS1, TDH, cSDH
Chromosome Location:
Chr.12: 113422265 - 113438277 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05568456_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDSL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001304993.1 NP_001291922.1 8 1182 87
NM_138432.3 NP_612441.1 9 1304 87
XM_005253831.4 XP_005253888.1 8 1181 87
XM_011537846.2 XP_011536148.1 8 1717 87
XM_017018763.1 XP_016874252.1 8 1181 87
GenBank mRNA AF134473.1 - 8 87
BC009849.2 - 9 1283 4040570 87
BC091479.1 - 8 1159 30344219 87

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SDSL

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

cellular amino acid metabolic process
biological_process
molecular_function
L-serine ammonia-lyase activity
L-threonine ammonia-lyase activity
pyridoxal phosphate binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0