This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05577546_g1
See other ERVH48-1 GE Assays ›
Gene Symbol:
ERVH48-1
Gene Name:
endogenous retrovirus group 48 member 1
Gene Aliases:
C21orf105, NDUFV3-AS1
Chromosome Location:
Chr.21: 42917251 - 42925598 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05577546_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ERVH48-1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001308491.1 NP_001295420.1 2 834 100
GenBank mRNA AB610407.1 - 2 834 100
BC005107.1 - 2 854 100

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

syncytium formation
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-fcf5f9d6d-dcxfx:80/100.66.77.77:80.
git-commit: 856de8fa7eff2d55603d92afdf035972e45c3e8a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.51.0-2026.05.21-1.0