This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05588262_s1
See other COL25A1 GE Assays ›
Gene Symbol:
COL25A1
Gene Name:
collagen type XXV alpha 1
Gene Aliases:
AMY, CFEOM5, CLAC, CLAC-P, CLACP
Chromosome Location:
Chr.4: 108808725 - 109302737 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05588262_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COL25A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011532333.2 XP_011530635.1 40 7002 142
XM_011532334.2 XP_011530636.1 40 6868 142
XM_011532335.2 XP_011530637.1 40 6697 142
XM_011532338.2 XP_011530640.1 38 7077 142
XM_011532355.2 XP_011530657.1 39 10781 142
XM_011532356.2 XP_011530658.1 38 10763 142
XM_011532358.2 XP_011530660.1 35 6173 142
XM_017008735.1 XP_016864224.1 38 6644 142
XM_017008736.1 XP_016864225.1 37 6979 142
XM_017008737.1 XP_016864226.1 39 9458 142

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

collagen catabolic process
axonogenesis involved in innervation
beta-amyloid binding
heparin binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline