Assay Details
| Gene Symbol: FGFRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004356.2 | NP_001004356.1 | 7 | 2995 | 107 | |
| NM_001004358.1 | NP_001004358.1 | 7 | 2878 | 107 | ||
| NM_021923.3 | NP_068742.2 | 6 | 2868 | 107 | ||
| XM_011513486.1 | XP_011511788.1 | 7 | 2999 | 107 | ||
| GenBank mRNA | AF312678.1 | - | 3179 | 107 | ||
| AJ277437.1 | - | 6 | 2868 | 107 | ||
| AK000530.1 | - | 1 | 533 | 107 | ||
| AK022597.1 | - | 1 | 1780 | 107 | ||
| AV707425.1 | - | 1 | 259 | 107 | ||
| AY358303.1 | - | 3200 | 107 | |||
| BC015228.1 | - | 3 | 2279 | 107 | ||
| BC036769.1 | - | 7 | 3007 | 5744684 | 107 | |
More Information
Set Membership: |
|

FGFRL1
Genomic Map