This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05594049_s1
See other NME7 GE Assays ›
Gene Symbol:
NME7
Gene Name:
NME/NM23 family member 7
Gene Aliases:
CFAP67, MN23H7, NDK 7, NDK7, nm23-H7
Chromosome Location:
Chr.1: 169132385 - 169367963 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05594049_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NME7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017001099.1 XP_016856588.1 12 2879 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

kinase transferase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

nucleoside diphosphate phosphorylation
GTP biosynthetic process
UTP biosynthetic process
CTP biosynthetic process
nucleoside diphosphate kinase activity
protein binding
ATP binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-fvx7n:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline