This product has been added to your favorites list. Go to
My Favorites
Please enter the information below and press OK to send your cart to Core Services for purchase.
Name
Phone number
Email
Budget number
PI Name
PI Email
Building and Room number
Pickup
Delivery
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Hs05607619_s1
See other HECW1 GE Assays ›
Gene Symbol:
HECW1
Gene Name:
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 1
Gene Aliases:
NEDL1
Chromosome Location:
Chr.7: 43112580 - 43566001 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID
Hs05607619_s1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
4,430.00
Your Price
Online offer:
4430.0
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: HECW1
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001287059.1
NP_001273988.1
10
1625
126
NM_015052.4
NP_055867.3
11
1860
126
XM_005249665.3
XP_005249722.1
10
1642
126
XM_006715670.3
XP_006715733.1
9
1540
126
XM_006715671.3
XP_006715734.1
9
1537
126
XM_006715673.3
XP_006715736.1
9
1537
126
XM_011515220.2
XP_011513522.1
10
1433
126
XM_011515222.2
XP_011513524.1
9
1574
126
XM_011515223.2
XP_011513525.1
9
1375
126
XM_011515224.2
XP_011513526.1
9
1855
126
XM_011515225.2
XP_011513527.2
7
892
126
XM_011515226.2
XP_011513528.1
6
1116
126
XM_017011882.1
XP_016867371.1
9
1882
126
XM_017011883.1
XP_016867372.1
10
1819
126
XM_017011884.1
XP_016867373.1
9
1879
126
XM_017011885.1
XP_016867374.1
11
1815
126
XM_017011886.1
XP_016867375.1
10
1644
126
XM_017011887.1
XP_016867376.1
9
1882
126
XM_017011888.1
XP_016867377.1
9
1879
126
XM_017011889.1
XP_016867378.1
10
1936
126
XM_017011890.1
XP_016867379.1
10
1644
126
GenBank mRNA
AB002320.2
-
11
1860
126
AB048365.1
-
8
1212
126
AK294918.1
-
10
1579
126
AL079731.1
-
1
126
BC151227.1
-
11
1860
9007236
126
Back To Top
More Information
Set Membership:
Ambion siRNA available
Amplicon length greater than or equal to 101
Mammalian Gene Collection
Detect Genomic DNA
Homo sapiens
Both primers and probe map within a single exon
Matching Ambion siRNAs:
Click here to find siRNAs for Gene HECW1
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
negative regulation of canonical Wnt signaling pathway
ubiquitin-protein transferase activity
ligase activity
ubiquitin protein ligase activity
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0