This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05616532_s1
See other PTPN3 GE Assays ›
Gene Symbol:
PTPN3
Gene Name:
protein tyrosine phosphatase, non-receptor type 3
Gene Aliases:
PTP-H1, PTPH1
Chromosome Location:
Chr.9: 109375694 - 109538395 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05616532_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPN3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145368.1 NP_001138840.1 25 5937 95
NM_001145369.1 NP_001138841.1 21 8121 95
NM_001145370.1 NP_001138842.1 20 7986 95
NM_001145371.1 NP_001138843.1 16 5555 95
NM_001145372.1 NP_001138844.1 15 5420 95
NM_002829.3 NP_002820.3 26 6072 95
NR_026918.1 13 4842 95
XM_006717197.3 XP_006717260.1 26 6181 95
XM_006717199.3 XP_006717262.1 26 6206 95
XM_006717201.3 XP_006717264.1 25 6013 95
XM_006717202.3 XP_006717265.1 23 5775 95
XM_006717203.3 XP_006717266.1 23 5734 95
XM_006717204.3 XP_006717267.1 26 6108 95
XM_011518888.2 XP_011517190.1 26 6142 95
XM_011518889.2 XP_011517191.1 26 6812 95
XM_017014955.1 XP_016870444.1 26 6164 95
XM_017014956.1 XP_016870445.1 24 5885 95
XM_017014957.1 XP_016870446.1 25 5973 95
GenBank mRNA AK096975.1 - 1 1737 95
CR749204.1 - 14 4993 95

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PTPN3

Gene Ontology Categories:

Function(s) Process(es)

protein dephosphorylation
peptidyl-tyrosine dephosphorylation
negative regulation of mitotic cell cycle
negative regulation of membrane protein ectodomain proteolysis
liver regeneration
regulation of membrane depolarization during action potential
regulation of sodium ion transmembrane transporter activity
phosphotyrosine binding
protein tyrosine phosphatase activity
protein binding
cytoskeletal protein binding
sodium channel regulator activity
ATPase binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline