This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05618354_s1
See other AUH GE Assays ›
Gene Symbol:
AUH
Gene Name:
AU RNA binding protein/enoyl-CoA hydratase
Gene Aliases:
-
Chromosome Location:
Chr.9: 91213815 - 91361969 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05618354_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AUH
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005252072.2 XP_005252129.1 7 6461 108

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

hydratase

Gene Ontology Categories:

Function(s) Process(es)

leucine catabolic process
fatty acid beta-oxidation
branched-chain amino acid catabolic process
mRNA 3'-UTR binding
enoyl-CoA hydratase activity
methylglutaconyl-CoA hydratase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0