This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05623074_s1
See other ACE2 GE Assays ›
Gene Symbol:
ACE2
Gene Name:
angiotensin I converting enzyme 2
Gene Aliases:
ACEH
Chromosome Location:
Chr.X: 15494525 - 15602069 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05623074_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACE2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011545549.1 XP_011543851.1 18 3060 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

regulation of cytokine production
angiotensin maturation
angiotensin catabolic process in blood
angiotensin-mediated drinking behavior
regulation of systemic arterial blood pressure by renin-angiotensin
tryptophan transport
regulation of vasoconstriction
receptor biosynthetic process
regulation of cell proliferation
regulation of vasodilation
viral entry into host cell
receptor-mediated virion attachment to host cell
regulation of inflammatory response
positive regulation of cardiac muscle contraction
positive regulation of gap junction assembly
regulation of cardiac conduction
positive regulation of reactive oxygen species metabolic process
virus receptor activity
glycoprotein binding
endopeptidase activity
carboxypeptidase activity
metallocarboxypeptidase activity
protein binding
metallopeptidase activity
peptidyl-dipeptidase activity
zinc ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0