This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05625961_s1
See other STAM GE Assays ›
Gene Symbol:
STAM
Gene Name:
signal transducing adaptor molecule
Gene Aliases:
STAM-1, STAM1
Chromosome Location:
Chr.10: 17644125 - 17716824 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05625961_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STAM
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001324282.1 NP_001311211.1 13 2942 126
NM_001324283.1 NP_001311212.1 13 2953 126
NM_001324284.1 NP_001311213.1 15 3142 126
NM_001324285.1 NP_001311214.1 15 3150 126
NM_001324286.1 NP_001311215.1 11 2781 126
NM_001324287.1 NP_001311216.1 12 2866 126
NM_001324288.1 NP_001311217.1 15 3181 126
NM_001324289.1 NP_001311218.1 14 3096 126
NM_003473.3 NP_003464.1 14 3038 126

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene STAM

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

intracellular protein transport
autophagy
signal transduction
positive regulation of signal transduction
endosomal transport
multivesicular body assembly
negative regulation of epidermal growth factor receptor signaling pathway
positive regulation of exosomal secretion
regulation of extracellular exosome assembly
SH3/SH2 adaptor activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline