This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05627034_s1
See other P2RX7 GE Assays ›
Gene Symbol:
P2RX7
Gene Name:
purinergic receptor P2X 7
Gene Aliases:
P2X7
Chromosome Location:
Chr.12: 121132819 - 121189478 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05627034_s1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: P2RX7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011538419.2 XP_011536721.1 12 3541 90
XM_011538420.2 XP_011536722.1 11 3208 90
XM_017019364.1 XP_016874853.1 13 3854 90
XM_017019365.1 XP_016874854.1 12 3582 90
XM_017019366.1 XP_016874855.1 12 3569 90
XM_017019367.1 XP_016874856.1 11 3407 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

ligand-gated ion channel

Gene Ontology Categories:

Function(s) Process(es)

activation of MAPK activity
cell morphogenesis
phagolysosome assembly
positive regulation of T cell mediated cytotoxicity
regulation of sodium ion transport
protein phosphorylation
membrane protein ectodomain proteolysis
phospholipid transfer to membrane
membrane budding
inflammatory response
mitochondrion organization
cell surface receptor signaling pathway
blood coagulation
response to mechanical stimulus
response to zinc ion
positive regulation of calcium ion transport into cytosol
positive regulation of gene expression
positive regulation of glutamate secretion
positive regulation of gamma-aminobutyric acid secretion
synaptic vesicle exocytosis
protein processing
phospholipid scrambling
sensory perception of pain
cytolysis
positive regulation of bone mineralization
cellular response to extracellular stimulus
bleb assembly
positive regulation of prostaglandin secretion
response to lipopolysaccharide
positive regulation of interleukin-6 production
collagen metabolic process
response to ATP
response to fluid shear stress
positive regulation of ion transmembrane transport
purinergic nucleotide receptor signaling pathway
T cell proliferation
response to drug
T cell homeostasis
NAD transport
negative regulation of MAPK cascade
multicellular organismal protein catabolic process
phospholipid translocation
negative regulation of bone resorption
negative regulation of cell volume
positive regulation of glycolytic process
positive regulation of cytolysis
ceramide biosynthetic process
pore complex assembly
skeletal system morphogenesis
homeostasis of number of cells within a tissue
positive regulation of interleukin-1 alpha secretion
positive regulation of interleukin-1 beta secretion
defense response to Gram-positive bacterium
release of sequestered calcium ion into cytosol
protein oligomerization
positive regulation of cytoskeleton organization
response to calcium ion
response to electrical stimulus
regulation of killing of cells of other organism
membrane depolarization
positive regulation of mitochondrial depolarization
positive regulation of lymphocyte apoptotic process
cellular response to dsRNA
reactive oxygen species metabolic process
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
positive regulation of bleb assembly
lipopolysaccharide binding
purinergic nucleotide receptor activity
extracellular ATP-gated cation channel activity
receptor binding
protein binding
ATP binding
protein homodimerization activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0