This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05627549_s1
See other SLC7A1 GE Assays ›
Gene Symbol:
SLC7A1
Gene Name:
solute carrier family 7 member 1
Gene Aliases:
ATRC1, CAT-1, ERR, HCAT1, REC1L
Chromosome Location:
Chr.13: 29509410 - 29595715 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05627549_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC7A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003045.4 NP_003036.1 13 5177 96
XM_017020713.1 XP_016876202.1 13 5244 96
XM_017020714.1 XP_016876203.1 13 5183 96
XM_017020715.1 XP_016876204.1 13 5065 96
GenBank mRNA AB105377.1 - 13 5064 96
AK124591.1 - 1 2497 96
AL050021.1 - 1 919 96

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC7A1

Panther Classification:

Molecular Function -

amino acid transporter

Gene Ontology Categories:

Function(s) Process(es)

transport
amino acid transport
arginine transport
L-ornithine transmembrane transport
L-lysine transmembrane transport
arginine transmembrane transport
L-ornithine transmembrane transporter activity
protein binding
amino acid transmembrane transporter activity
arginine transmembrane transporter activity
L-lysine transmembrane transporter activity
antiporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0