This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05629161_s1
See other MXRA8 GE Assays ›
Gene Symbol:
MXRA8
Gene Name:
matrix remodeling associated 8
Gene Aliases:
-
Chromosome Location:
Chr.1: 1352689 - 1363541 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05629161_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MXRA8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282582.1 NP_001269511.1 6 848 109
NM_001282583.1 NP_001269512.1 5 1267 109
NM_001282584.1 NP_001269513.1 4 767 109
NM_001282585.1 NP_001269514.1 5 823 109
NM_032348.3 NP_115724.1 5 1070 109
XM_017001516.1 XP_016857005.1 5 847 109
XM_017001517.1 XP_016857006.1 4 2474 109
GenBank mRNA AB052096.1 - 6 848 109
AK027269.1 - 5 823 109
AK095966.1 - 5 1267 109
AK225028.1 - 5 809 109
AK293442.1 - 4 536 109
AK304615.1 - 5 741 109
AK315510.1 - 5 837 109
AY358391.1 - 4 513 109
BC006213.2 - 5 811 3343261 109
BC017312.2 - 5 811 5090754 109

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MXRA8

Gene Ontology Categories:

Function(s) Process(es)

establishment of glial blood-brain barrier
molecular_function

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0