This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05638611_s1
See other TMED3 GE Assays ›
Gene Symbol:
TMED3
Gene Name:
transmembrane p24 trafficking protein 3
Gene Aliases:
C15orf22, P24B, p24g4, p26
Chromosome Location:
Chr.15: 79311062 - 79419879 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05638611_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMED3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017022040.1 XP_016877529.1 3 6616 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s)

ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
protein transport

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0