This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05639726_s1
See other NQO1 GE Assays ›
Gene Symbol:
NQO1
Gene Name:
NAD(P)H quinone dehydrogenase 1
Gene Aliases:
DHQU, DIA4, DTD, NMOR1, NMORI, QR1
Chromosome Location:
Chr.16: 69709401 - 69726668 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05639726_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NQO1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000903.2 NP_000894.1 6 1558 74
NM_001025433.1 NP_001020604.1 5 1456 74
NM_001025434.1 NP_001020605.1 5 1444 74
NM_001286137.1 NP_001273066.1 4 1380 74
GenBank mRNA BM052991.1 - 1 485 74
BM787983.1 - 1 191 74
J03934.1 - 6 1417 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NQO1

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

regulation of cellular amino acid metabolic process
xenobiotic metabolic process
nitric oxide biosynthetic process
synaptic transmission, cholinergic
aging
response to nutrient
response to toxic substance
removal of superoxide radicals
response to estradiol
negative regulation of catalytic activity
positive regulation of neuron apoptotic process
response to ethanol
oxidation-reduction process
NAD(P)H dehydrogenase (quinone) activity
cytochrome-b5 reductase activity, acting on NAD(P)H
superoxide dismutase activity
protein binding
identical protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0