This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05641794_s1
See other ABR GE Assays ›
Gene Symbol:
ABR
Gene Name:
active BCR-related
Gene Aliases:
MDB
Chromosome Location:
Chr.17: 1003518 - 1229713 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05641794_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ABR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011523811.2 XP_011522113.2 1 55 99
XM_017024539.1 XP_016880028.1 1 52 99
XM_017024540.1 XP_016880029.1 1 53 99
XM_017024541.1 XP_016880030.1 1 55 99

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011523811.2, XM_017024539.1, XM_017024540.1, XM_017024541.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

G-protein modulator

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of cellular extravasation
small GTPase mediated signal transduction
brain development
actin cytoskeleton organization
response to lipopolysaccharide
regulation of Rho protein signal transduction
inner ear morphogenesis
positive regulation of apoptotic process
regulation of vascular permeability
negative regulation of neutrophil degranulation
positive regulation of GTPase activity
negative regulation of inflammatory response
positive regulation of phagocytosis
neuromuscular process controlling balance
regulation of small GTPase mediated signal transduction
negative regulation of blood vessel remodeling
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
GTPase activator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4g6l8:80/100.66.76.118:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0