This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05642305_s1
See other ALDOC GE Assays ›
Gene Symbol:
ALDOC
Gene Name:
aldolase, fructose-bisphosphate C
Gene Aliases:
ALDC
Chromosome Location:
Chr.17: 28573115 - 28576963 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05642305_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDOC
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005165.2 NP_005156.1 9 1529 63
XM_005257949.2 XP_005258006.1 10 1615 63
XM_011524556.1 XP_011522858.1 10 1544 63
GenBank mRNA AF054987.1 - 9 1440 63
AK293623.1 - 9 1369 63
AK293693.1 - 8 1374 63
AK294157.1 - 9 1422 63
BC003613.2 - 9 1475 2987869 63
BC065565.1 - 9 1475 6155379 63
BC103760.1 - 9 1487 63
BC106925.1 - 9 1440 40032967 63
BC106926.1 - 9 1440 40032968 63
BF345067.1 - 1 36 63

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ALDOC

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

fructose metabolic process
gluconeogenesis
fructose 1,6-bisphosphate metabolic process
epithelial cell differentiation
canonical glycolysis
fructose-bisphosphate aldolase activity
protein binding
cytoskeletal protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0