This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05644820_s1
See other PYCR1 GE Assays ›
Gene Symbol:
PYCR1
Gene Name:
pyrroline-5-carboxylate reductase 1
Gene Aliases:
ARCL2B, ARCL3B, P5C, P5CR, PIG45, PP222, PRO3, PYCR
Chromosome Location:
Chr.17: 81932384 - 81937328 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05644820_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PYCR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282279.1 NP_001269208.1 7 1577 108
NM_001282280.1 NP_001269209.1 8 1670 108
NM_001282281.1 NP_001269210.1 8 1663 108
NM_006907.3 NP_008838.2 7 1999 108
NM_153824.2 NP_722546.1 8 1656 108
XM_005256381.2 XP_005256438.1 8 1628 108
XM_011523583.2 XP_011521885.1 8 1867 108
XM_011523584.2 XP_011521886.1 8 1622 108
XM_011523585.2 XP_011521887.1 7 1493 108
XM_017024909.1 XP_016880398.1 7 1506 108
GenBank mRNA AA026075.1 - 1 108
AF218000.1 - 8 1492 108
AK225879.1 - 7 1545 108
BC001504.2 - 7 1596 3505512 108
BC022244.1 - 7 1517 4420238 108
BC071842.1 - 7 1618 5088067 108
M77836.1 - 7 1567 108

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PYCR1

Panther Classification:

Molecular Function -

reductase

Gene Ontology Categories:

Function(s) Process(es)

proline biosynthetic process
cellular amino acid biosynthetic process
cellular response to oxidative stress
regulation of mitochondrial membrane potential
oxidation-reduction process
L-proline biosynthetic process
negative regulation of hydrogen peroxide-induced cell death
pyrroline-5-carboxylate reductase activity
protein binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mmql6:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0