This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05645071_s1
See other POLR2A GE Assays ›
Gene Symbol:
POLR2A
Gene Name:
RNA polymerase II subunit A
Gene Aliases:
POLR2, POLRA, RPB1, RPBh1, RPO2, RPOL2, RpIILS, hRPB220, hsRPB1
Chromosome Location:
Chr.17: 7484379 - 7514618 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05645071_s1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: POLR2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000937.4 NP_000928.1 29 6502 113
GenBank mRNA AK304882.1 - 28 6148 113
AW589665.1 - 1 237 113
BU730469.1 - 1 249 113
X63564.1 - 29 6502 113

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Inventoried Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene POLR2A

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

mRNA splicing, via spliceosome
transcription, RNA-templated
transcription-coupled nucleotide-excision repair
DNA-templated transcription, termination
regulation of transcription, DNA-templated
transcription from RNA polymerase II promoter
transcription initiation from RNA polymerase II promoter
transcription elongation from RNA polymerase II promoter
7-methylguanosine mRNA capping
fibroblast growth factor receptor signaling pathway
gene expression
gene silencing by RNA
positive regulation of RNA splicing
somatic stem cell population maintenance
snRNA transcription from RNA polymerase II promoter
positive regulation of viral transcription
RNA polymerase II activity
DNA binding
DNA-directed RNA polymerase activity
RNA-directed RNA polymerase activity
protein binding
ubiquitin protein ligase binding
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0