This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05645100_s1
See other ARHGAP44 GE Assays ›
Gene Symbol:
ARHGAP44
Gene Name:
Rho GTPase activating protein 44
Gene Aliases:
NPC-A-10, RICH2
Chromosome Location:
Chr.17: 12789512 - 12991644 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05645100_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP44
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321164.1 NP_001308093.1 21 2912 63
NM_001321166.1 NP_001308095.1 20 2860 63
NM_001321167.1 NP_001308096.1 22 2930 63
NM_001321168.1 NP_001308097.1 20 2794 63
NM_014859.5 NP_055674.4 21 2878 63
NR_135569.1 18 2462 63
XM_005256891.1 XP_005256948.1 19 2488 63
GenBank mRNA AB014572.1 - 21 2851 63
AL080100.1 - 1 144 63
AL096728.1 - 9 1522 63
BC022452.1 - 3 632 63
BC070074.1 - 20 2572 63
BC094754.1 - 20 2572 63
BC117412.1 - 21 2630 40125963 63
BC117416.1 - 21 2630 40125967 63
BC143852.1 - 22 2682 63
BC143853.1 - 20 2612 9052369 63
BC143855.1 - 18 2214 63
BC143856.1 - 21 2664 63

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ARHGAP44

Gene Ontology Categories:

Function(s) Process(es)

exocytosis
signal transduction
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
GTPase activator activity
protein binding
phospholipid binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-r7bw6:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0