This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05646089_s1
See other ACE GE Assays ›
Gene Symbol:
ACE
Gene Name:
angiotensin I converting enzyme
Gene Aliases:
ACE1, CD143, DCP, DCP1, ICH, MVCD3
Chromosome Location:
Chr.17: 63477061 - 63498380 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05646089_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ACE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000789.3 NP_000780.1 25 4530 101
NM_001178057.1 NP_001171528.1 13 2702 101
NM_152830.2 NP_690043.1 14 2825 101
GenBank mRNA JQ927553.1 - 24 4681 101

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ACE

Panther Classification:

Molecular Function -

metalloprotease

Gene Ontology Categories:

Function(s) Process(es)

kidney development
blood vessel remodeling
angiotensin maturation
angiotensin catabolic process in blood
regulation of renal output by angiotensin
neutrophil mediated immunity
antigen processing and presentation of peptide antigen via MHC class I
regulation of systemic arterial blood pressure by renin-angiotensin
spermatogenesis
regulation of blood pressure
regulation of smooth muscle cell migration
regulation of vasoconstriction
mononuclear cell proliferation
regulation of vasodilation
hormone catabolic process
peptide catabolic process
beta-amyloid metabolic process
arachidonic acid secretion
heart contraction
regulation of angiotensin metabolic process
hematopoietic stem cell differentiation
positive regulation of protein tyrosine kinase activity
cell proliferation in bone marrow
positive regulation of peptidyl-tyrosine autophosphorylation
regulation of hematopoietic stem cell proliferation
negative regulation of gap junction assembly
positive regulation of peptidyl-cysteine S-nitrosylation
endopeptidase activity
carboxypeptidase activity
drug binding
metallopeptidase activity
exopeptidase activity
tripeptidyl-peptidase activity
peptidyl-dipeptidase activity
zinc ion binding
chloride ion binding
mitogen-activated protein kinase kinase binding
bradykinin receptor binding
mitogen-activated protein kinase binding
metallodipeptidase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0