This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05658694_s1
See other LCLAT1 GE Assays ›
Gene Symbol:
LCLAT1
Gene Name:
lysocardiolipin acyltransferase 1
Gene Aliases:
1AGPAT8, AGPAT8, ALCAT1, HSRG1849, LYCAT, UNQ1849
Chromosome Location:
Chr.2: 30447226 - 30644225 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05658694_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LCLAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002257.2 NP_001002257.1 6 1594 89
NM_001304445.1 NP_001291374.1 8 1999 89
NM_001304446.1 NP_001291375.1 6 1603 89
NM_182551.4 NP_872357.2 7 1786 89
XM_005264244.1 XP_005264301.1 7 1748 89
XM_005264245.3 XP_005264302.1 6 1600 89
XM_011532741.2 XP_011531043.1 7 1737 89
XM_011532742.2 XP_011531044.1 7 1564 89
XM_011532743.2 XP_011531045.1 6 1551 89
XM_011532744.2 XP_011531046.1 7 1746 89
XM_017003746.1 XP_016859235.1 7 1631 89
GenBank mRNA AK311294.1 - 2 572 89
BC146817.1 - 6 1584 8860419 89
BX647953.1 - 6 1551 89

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LCLAT1

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

phosphatidic acid biosynthetic process
multicellular organism development
CDP-diacylglycerol biosynthetic process
triglyceride biosynthetic process
cardiolipin acyl-chain remodeling
1-acylglycerol-3-phosphate O-acyltransferase activity
protein binding
O-acyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0