This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs05661901_s1
See other ZNF343 GE Assays ›
Gene Symbol:
ZNF343
Gene Name:
zinc finger protein 343
Gene Aliases:
dJ734P14.5
Chromosome Location:
Chr.20: 2481817 - 2524522 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs05661901_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ZNF343
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001282495.1 NP_001269424.1 7 2292 81
NM_001282496.1 NP_001269425.1 6 2324 81
NM_001282497.1 NP_001269426.1 7 2489 81
NM_001282498.1 NP_001269427.1 4 2040 81
NM_001321800.1 NP_001308729.1 6 2230 81
NM_001321801.1 NP_001308730.1 7 2353 81
NM_001321802.1 NP_001308731.1 7 2486 81
NM_001321803.1 NP_001308732.1 7 2350 81
NM_001321805.1 NP_001308734.1 6 2363 81
NM_024325.5 NP_077301.4 6 2366 81
XM_017028062.1 XP_016883551.1 6 2207 81
GenBank mRNA AK055734.1 - 2 1795 81
AK096911.1 - 6 2367 81
BC011862.2 - 1 1536 81
BC065009.1 - 7 2289 6166526 81
BQ212757.1 - 249 81

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ZNF343

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline