This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06012631_m1
See other PCDH1 GE Assays ›
Gene Symbol:
PCDH1
Gene Name:
protocadherin 1
Gene Aliases:
PC42, PCDH42
Chromosome Location:
Chr.5: 141852319 - 141878410 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06012631_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCDH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278613.1 NP_001265542.1 2 - 3 973 93
NM_002587.4 NP_002578.2 2 - 3 1050 93
NM_032420.3 NP_115796.2 2 - 3 1050 93
XM_005268452.2 XP_005268509.1 2 - 3 3380 93
XM_005268454.4 XP_005268511.1 2 - 3 3380 93
GenBank mRNA AK075233.1 - 1 - 2 419 93
AK289512.1 - 2 - 3 1050 93
AK304231.1 - 2 - 3 935 93
BC035812.1 - 2 - 3 1019 5753628 93

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene PCDH1

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
cell-cell signaling
nervous system development
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0