This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06060855_m1
See other MIER1 GE Assays ›
Gene Symbol:
MIER1
Gene Name:
MIER1 transcriptional regulator
Gene Aliases:
ER1, MI-ER1
Chromosome Location:
Chr.1: 66924895 - 66988619 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06060855_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MIER1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077702.2 NP_001071170.2 3 - 4 374 71
NM_001146110.1 NP_001139582.1 3 - 4 377 71
NM_001146111.1 NP_001139583.1 3 - 4 374 71
NM_001278215.1 NP_001265144.1 3 - 4 374 71
NM_020948.3 NP_065999.2 3 - 4 374 71
XM_005271076.3 XP_005271133.1 3 - 4 412 71
XM_017001924.1 XP_016857413.1 3 - 4 268 71
XM_017001925.1 XP_016857414.1 3 - 4 373 71
XM_017001926.1 XP_016857415.1 4 - 5 312 71
GenBank mRNA AF515448.2 - 3 - 4 311 71
AL831987.2 - 3 - 4 316 71
AY124186.2 - 3 - 4 311 71
AY124189.2 - 3 - 4 311 71
AY124192.2 - 3 - 4 311 71
BC108726.1 - 3 - 4 255 5194296 71

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MIER1

Panther Classification:

Molecular Function -

homeodomain transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
signal transduction
positive regulation of chromatin silencing
positive regulation of I-kappaB kinase/NF-kappaB signaling
DNA binding
signal transducer activity
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zsvtd:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0