This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06596631_m1
See other DCLRE1C GE Assays ›
Gene Symbol:
DCLRE1C
Gene Name:
DNA cross-link repair 1C
Gene Aliases:
A-SCID, DCLREC1C, RS-SCID, SCIDA, SNM1C
Chromosome Location:
Chr.10: 14897853 - 14954432 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06596631_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DCLRE1C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033855.2 NP_001029027.1 12 - 13 1486 94
NM_001033857.2 NP_001029029.1 13 - 14 1571 94
NM_001033858.2 NP_001029030.1 14 - 15 1893 94
NM_001289076.1 NP_001276005.1 10 - 11 1345 94
NM_001289077.1 NP_001276006.1 13 - 14 1617 94
NM_001289078.1 NP_001276007.1 10 - 11 1378 94
NM_001289079.1 NP_001276008.1 14 - 15 1939 94
NM_022487.3 NP_071932.2 11 - 12 1430 94
NR_110297.1 14 - 15 1698 94
XM_006717491.3 XP_006717554.1 12 - 13 1160 94
XM_011519616.1 XP_011517918.1 12 - 13 1176 94
XM_011519617.1 XP_011517919.1 13 - 14 1295 94
XM_011519619.1 XP_011517921.1 14 - 15 1322 94
XM_011519620.2 XP_011517922.1 12 - 13 1161 94
XM_017016556.1 XP_016872045.1 12 - 13 1161 94
XM_017016557.1 XP_016872046.1 12 - 13 1188 94
XM_017016558.1 XP_016872047.1 13 - 14 1234 94
XM_017016559.1 XP_016872048.1 10 - 11 1029 94
XM_017016560.1 XP_016872049.1 13 - 14 1250 94
GenBank mRNA AF395747.1 - 13 - 14 1187 94
AF395748.1 - 13 - 14 1233 94
AF395749.1 - 11 - 12 1046 94
AF395750.1 - 10 - 11 994 94
AF395751.1 - 14 - 15 1509 94
AF395752.1 - 14 - 15 1555 94
AJ296101.1 - 12 - 13 1102 94
AK021422.1 - 11 - 12 1093 94
AK022922.1 - 14 - 15 1492 94
AK093757.1 - 1 - 2 699 94
AK290598.1 - 12 - 13 1149 94
BC009185.2 - 10 - 11 969 3945623 94
BC022254.1 - 12 - 13 1086 4272155 94
BX648858.1 - 13 - 14 1203 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK093757.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DCLRE1C

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
double-strand break repair via nonhomologous end joining
response to ionizing radiation
B cell differentiation
protection from non-homologous end joining at telomere
V(D)J recombination
interstrand cross-link repair
nucleic acid phosphodiester bond hydrolysis
single-stranded DNA endodeoxyribonuclease activity
damaged DNA binding
endonuclease activity
protein binding
5'-3' exonuclease activity
5'-3' exodeoxyribonuclease activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0