This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06596636_m1
See other MGMT GE Assays ›
Gene Symbol:
MGMT
Gene Name:
O-6-methylguanine-DNA methyltransferase
Gene Aliases:
-
Chromosome Location:
Chr.10: 129467184 - 129770983 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06596636_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MGMT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002412.4 NP_002403.2 1 - 2 109 74
GenBank mRNA AK293593.1 - 1 - 2 79 74
BC000824.1 - 1 - 2 37 3454207 74
BI520029.1 - 1 - 2 91 74
DC387686.1 - 1 - 2 103 74
DC416206.1 - 1 - 2 109 74
M29971.1 - 1 - 2 80 74

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC000824.1, M29971.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MGMT

Gene Ontology Categories:

Function(s) Process(es)

DNA ligation
DNA methylation
DNA dealkylation involved in DNA repair
response to toxic substance
cellular response to oxidative stress
response to drug
negative regulation of apoptotic process
regulation of cysteine-type endopeptidase activity involved in apoptotic process
response to ethanol
response to folic acid
mammary gland epithelial cell differentiation
cellular response to organic cyclic compound
cellular response to ionizing radiation
positive regulation of double-strand break repair
DNA binding
damaged DNA binding
methylated-DNA-[protein]-cysteine S-methyltransferase activity
calcium ion binding
methyltransferase activity
DNA-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0