This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06597688_m1
See other CDH18 GE Assays ›
Gene Symbol:
CDH18
Gene Name:
cadherin 18
Gene Aliases:
CDH14, CDH14L, CDH24
Chromosome Location:
Chr.5: 19472930 - 20575902 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06597688_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDH18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005248228.3 XP_005248285.1 2 - 3 94
XM_017008924.1 XP_016864413.1 3 - 4 612 213
XM_017008925.1 XP_016864414.1 3 - 4 608 209
XM_017008926.1 XP_016864415.1 3 - 4 469 213
XM_017008927.1 XP_016864416.1 3 - 4 398 213
XM_017008928.1 XP_016864417.1 4 - 5 656 213
XM_017008932.1 XP_016864421.1 3 - 4 331 209
XM_017008934.1 XP_016864423.1 2 - 3 94
XM_017008935.1 XP_016864424.1 3 - 4 612 213
XM_017008936.1 XP_016864425.1 2 - 3 94
XM_017008938.1 XP_016864427.1 2 - 3 94
XM_017008939.1 XP_016864428.1 3 - 4 589 213
XM_017008940.1 XP_016864429.1 2 - 3 94

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005248228.3, XM_017008924.1, XM_017008925.1, XM_017008926.1, XM_017008927.1, XM_017008928.1, XM_017008932.1, XM_017008934.1, XM_017008935.1, XM_017008936.1, XM_017008938.1, XM_017008939.1, XM_017008940.1 transcripts.

Set Membership:

Amplicon length between 86 and 100 Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
adherens junction organization
calcium ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0