This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06598023_mH
See other CDKN2B-AS1 GE Assays ›
Gene Symbol:
CDKN2B-AS1
Gene Name:
CDKN2B antisense RNA 1
Gene Aliases:
ANRIL, CDKN2B-AS, CDKN2BAS, NCRNA00089, PCAT12, p15AS
Chromosome Location:
Chr.9: 21994791 - 22121097 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06598023_mH
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CDKN2B-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_003529.3 6 - 7 1256 72
NR_047532.1 5 - 6 943 72
NR_047535.1 3 - 4 648 72
NR_047537.1 3 - 4 648 72
NR_047539.1 6 - 7 1256 72
NR_047540.1 3 - 4 648 72
NR_047541.1 3 - 4 648 72
NR_047542.1 3 - 4 648 72
NR_047543.1 3 - 4 648 72
GenBank mRNA AB548314.1 - 5 - 6 943 72
DQ485453.1 - 6 - 7 1255 72
DQ485454.1 - 6 - 7 1255 72
EU741058.1 - 3 - 4 344 72
GQ495921.1 - 3 - 4 537 72
GQ495922.1 - 3 - 4 537 72
GQ495923.1 - 3 - 4 537 72
GQ495924.1 - 3 - 4 537 72
GQ495926.1 - 3 - 4 648 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline