This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06598193_m1
See other ADGRL2 GE Assays ›
Gene Symbol:
ADGRL2
Gene Name:
adhesion G protein-coupled receptor L2
Gene Aliases:
CIRL2, CL2, LEC1, LPHH1, LPHN2
Chromosome Location:
Chr.1: 81306155 - 81992738 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06598193_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRL2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK123422.1 - 1 - 2 312 119
BC031321.1 - 1 - 2 356 119

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK123422.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0