This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06598201_m1
See other MOV10 GE Assays ›
Gene Symbol:
MOV10
Gene Name:
Mov10 RISC complex RNA helicase
Gene Aliases:
fSAP113, gb110
Chromosome Location:
Chr.1: 112674312 - 112702377 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06598201_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOV10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130079.2 NP_001123551.1 3 - 4 558 62
NM_001286072.1 NP_001273001.1 4 - 5 1582 62
NM_001321324.1 NP_001308253.1 3 - 4 459 62
NM_020963.4 NP_066014.1 3 - 4 834 62
XM_005270869.4 XP_005270926.1 4 - 5 785 62
XM_005270870.4 XP_005270927.1 4 - 5 801 62
XM_017001318.1 XP_016856807.1 4 - 5 1392 62
XM_017001319.1 XP_016856808.1 3 - 4 459 62
GenBank mRNA AB046851.1 - 4 - 5 1196 62
AK023297.1 - 3 - 4 436 62
AK074174.1 - 2 - 3 410 62
AK226011.1 - 3 - 4 461 62
AK301229.1 - 2 - 3 292 62
AK308874.1 - 3 - 4 470 62
AK310512.1 - 3 - 4 633 62
AL833353.1 - 3 - 4 725 62
BC002548.1 - 2 - 3 405 3138543 62
BC009312.2 - 2 - 3 429 4109453 62
BC025339.1 - 2 - 3 429 62
BX647787.1 - 4 - 5 1174 62

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MOV10

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
Wnt signaling pathway, calcium modulating pathway
gene silencing by miRNA
mRNA cleavage involved in gene silencing by miRNA
phosphatidylinositol-mediated signaling
helicase activity
protein binding
ATP binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-ng27p:80/100.66.78.145:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline