This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06599540_s1
See other TDG GE Assays ›
Gene Symbol:
TDG
Gene Name:
thymine DNA glycosylase
Gene Aliases:
hTDG
Chromosome Location:
Chr.12: 103965815 - 103988878 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06599540_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TDG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003211.4 NP_003202.3 2 376 100
XM_005269125.1 XP_005269182.1 2 236 100
GenBank mRNA AK299832.1 - 2 288 100
AK309792.1 - 3 420 100
AK313234.1 - 2 319 100
AL710869.1 - 3 528 100
BC019925.1 - 2 330 100
BC037557.1 - 2 329 5184772 100
BC104477.1 - 2 292 100
DB162106.1 - 2 377 100

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_005269125.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Amplicon spans exons and probe does not span exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TDG

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
base-excision repair
base-excision repair, AP site formation
mismatch repair
transcription, DNA-templated
embryo development
covalent chromatin modification
protein sumoylation
negative regulation of protein binding
oxidative DNA demethylation
negative regulation of chromatin binding
regulation of gene expression, epigenetic
depyrimidination
DNA demethylation
RNA polymerase II transcription cofactor activity
damaged DNA binding
double-stranded DNA binding
uracil DNA N-glycosylase activity
protein kinase C binding
protein binding
pyrimidine-specific mismatch base pair DNA N-glycosylase activity
DNA N-glycosylase activity
mismatched DNA binding
SUMO binding
protein homodimerization activity
structure-specific DNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline