This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06606883_g1
See other IGSF3 GE Assays ›
Gene Symbol:
IGSF3 GGT2
Gene Name:
immunoglobulin superfamily member 3
gamma-glutamyltransferase 2
Gene Aliases:
EWI-3, LCDD, V8
GGT, GGT 2
Chromosome Location:
Chr.1: 116574398 - 116667755 on Build GRCh38
Chromosome Location:
Chr.22: 21207972 - 21282982 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs06606883_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IGSF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005270794.4 XP_005270851.1 1 - 2 78 62
XM_006710593.3 XP_006710656.1 1 - 2 81 62
Gene Symbol: GGT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017029180.1 XP_016884669.1 1 - 2 223 62

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017029180.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily protease

Gene Ontology Categories:

Function(s) Process(es)

lacrimal gland development
glutathione metabolic process
glutathione biosynthetic process
xenobiotic metabolic process
leukotriene biosynthetic process
zymogen activation
molecular_function
gamma-glutamyltransferase activity
glutathione hydrolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0