This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06617526_s1
See other CHD1L GE Assays ›
Gene Symbol:
CHD1L
Gene Name:
chromodomain helicase DNA binding protein 1 like
Gene Aliases:
ALC1, CHDL
Chromosome Location:
Chr.1: 147172804 - 147295766 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06617526_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHD1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_017002858.1 XP_016858347.1 1 633 124

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_017002858.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
chromatin remodeling
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
global genome nucleotide-excision repair
nucleotide binding
nucleic acid binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
ATPase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0