This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06625872_s1
See other CABLES1 GE Assays ›
Gene Symbol:
CABLES1
Gene Name:
Cdk5 and Abl enzyme substrate 1
Gene Aliases:
CABL1, CABLES, HsT2563, IK3-1
Chromosome Location:
Chr.18: 23134564 - 23260470 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06625872_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CABLES1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001100619.2 NP_001094089.1 1 708 56
GenBank mRNA AF348525.1 - 1 108 56
EF028204.1 - 1 708 56

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Gene Ontology Categories:

Function(s) Process(es)

cell cycle
cell division
regulation of cell cycle
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0