This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06627321_g1
See other CHAT GE Assays ›
Gene Symbol:
CHAT
Gene Name:
choline O-acetyltransferase
Gene Aliases:
CHOACTASE, CMS1A, CMS1A2, CMS6
Chromosome Location:
Chr.10: 49609095 - 49665104 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06627321_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHAT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_020984.3 NP_066264.3 1 30 90
GenBank mRNA AF305909.1 - 1 30 90
D82342.1 - 1 30 90

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AF305909.1, D82342.1, NM_020984.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CHAT

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

antral ovarian follicle growth
response to hypoxia
phosphatidylcholine biosynthetic process
neurotransmitter secretion
response to nutrient
memory
acetylcholine biosynthetic process
response to drug
response to ethanol
choline O-acetyltransferase activity
choline binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0