This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06627846_gH
See other RPS9 GE Assays ›
Gene Symbol:
RPS9
Gene Name:
ribosomal protein S9
Gene Aliases:
S9
Chromosome Location:
Chr.19: 54200557 - 54207647 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06627846_gH
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RPS9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001013.3 NP_001004.2 363 69
NM_001321701.1 NP_001308630.1 4 368 69
NM_001321702.1 NP_001308631.1 4 400 69
NM_001321704.1 NP_001308633.1 4 357 69
NM_001321705.1 NP_001308634.1 3 240 69
NR_135762.1 4 368 69
NR_135763.1 4 363 69
GenBank mRNA AW163829.1 - 3 226 69
BC000802.1 - 3 338 3452221 69
BC007410.2 - 3 345 2964451 69
BC007434.2 - 3 345 2964451 69
BC007857.2 - 3 343 4298534 69
BC020462.1 - 2 69
BC068055.1 - 4 350 5454636 69
BC071940.1 - 3 345 6258045 69
BC071941.1 - 3 347 69
BC096756.1 - 3 350 3507400 69
BC096822.1 - 3 318 69
BC141920.1 - 2 124 69
BG753945.1 - 4 347 69
BG911482.1 - 4 355 69
BM313608.1 - 4 400 69
BQ069656.1 - 3 338 69
CD389050.1 - 4 390 69
U14971.1 - 3 342 69

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RPS9

Panther Classification:

Molecular Function -

ribosomal protein

Gene Ontology Categories:

Function(s) Process(es)

nuclear-transcribed mRNA catabolic process, nonsense-mediated decay
rRNA processing
translation
translational initiation
SRP-dependent cotranslational protein targeting to membrane
positive regulation of cell proliferation
viral transcription
positive regulation of translational fidelity
structural constituent of ribosome
protein binding
rRNA binding
poly(A) RNA binding
translation regulator activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline