This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06627962_s1
See other APBA3 GE Assays ›
Gene Symbol:
APBA3
Gene Name:
amyloid beta precursor protein binding family A member 3
Gene Aliases:
MGC:15815, X11L2, mint3
Chromosome Location:
Chr.19: 3750773 - 3761697 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06627962_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: APBA3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_006722950.3 XP_006723013.1 9 1793 70
XM_006722951.3 XP_006723014.1 7 1478 70
GenBank mRNA AB209603.1 - 3 3866 70
AF029110.1 - 1 71
BG938716.1 - 2 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

membrane trafficking regulatory protein

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
chemical synaptic transmission
regulation of gene expression
protein transport
negative regulation of catalytic activity
beta-amyloid binding
enzyme inhibitor activity
protein binding
enzyme binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0