This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06628244_s1
See other LILRB2 GE Assays ›
Gene Symbol:
LILRB2
Gene Name:
leukocyte immunoglobulin like receptor B2
Gene Aliases:
CD85D, ILT-4, ILT4, LIR-2, LIR2, MIR-10, MIR10
Chromosome Location:
Chr.19: 54273821 - 54281178 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06628244_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LILRB2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080978.3 NP_001074447.2 4 403 67
NM_001278403.2 NP_001265332.2 4 265 67
NM_001278405.2 NP_001265334.2 3 185 67
NM_001278406.2 NP_001265335.2 3 185 67
NM_005874.4 NP_005865.3 4 403 67
NR_103521.2 3 269 67
GenBank mRNA AF000574.1 - 3 137 67
AF004231.2 - 4 265 67
AF011565.1 - 3 139 67
AF011566.1 - 3 139 67
AF025528.1 - 4 320 67
AF283986.1 - 4 311 67
AF283987.1 - 3 186 67
AK226015.1 - 4 376 67
AK297071.1 - 4 358 67
AK298649.1 - 3 216 67
AK310263.1 - 3 215 67
BC036827.1 - 4 293 5212543 67
DC429031.1 - 3 269 67
DC429053.1 - 4 404 67
DQ655938.2 - 2 265 67
EU915606.1 - 3 137 67
LC125264.1 - 2 89 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LILRB2

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
negative regulation of antigen processing and presentation
positive regulation of tolerance induction
positive regulation of T cell tolerance induction
immune response-inhibiting cell surface receptor signaling pathway
Fc receptor mediated inhibitory signaling pathway
immune response
cellular defense response
signal transduction
cell surface receptor signaling pathway
cell-cell signaling
positive regulation of interleukin-6 production
heterotypic cell-cell adhesion
positive regulation of T cell proliferation
negative regulation of T cell proliferation
positive regulation of regulatory T cell differentiation
regulation of immune response
negative regulation of immune response
negative regulation of calcium ion transport
cellular response to lipopolysaccharide
regulation of dendritic cell differentiation
receptor activity
protein binding
protein phosphatase 1 binding
MHC class Ib protein binding
inhibitory MHC class I receptor activity
MHC class I protein binding
cell adhesion molecule binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0