This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06628887_s1
See other TAL1 GE Assays ›
Gene Symbol:
TAL1
Gene Name:
TAL bHLH transcription factor 1, erythroid differentiation factor
Gene Aliases:
SCL, TCL5, bHLHa17, tal-1
Chromosome Location:
Chr.1: 47216290 - 47232389 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06628887_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TAL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001287347.2 NP_001274276.1 2 172 74
NM_001290403.1 NP_001277332.1 2 282 74
NM_001290404.1 NP_001277333.1 3 287 74
NM_001290405.1 NP_001277334.1 2 155 74
NM_003189.5 NP_003180.1 3 400 74
XM_005271160.4 XP_005271217.1 2 1782 74
XM_017002187.1 XP_016857676.1 2 1197 74
XM_017002188.1 XP_016857677.1 2 389 74
XM_017002189.1 XP_016857678.1 2 192 74
XM_017002190.1 XP_016857679.1 2 888 74
XM_017002192.1 XP_016857681.1 2 888 74
XM_017002193.1 XP_016857682.1 2 196 74
GenBank mRNA M61108.1 - 1 533 74
S53245.1 - 2 172 74
S53686.1 - 2 167 74

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the M61108.1, NM_001287347.2, NM_001290403.1, NM_001290404.1, NM_001290405.1, NM_003189.5, S53245.1, XM_005271160.4, XM_017002187.1, XM_017002188.1, XM_017002189.1, XM_017002190.1, XM_017002193.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TAL1

Panther Classification:

Molecular Function -

basic helix-loop-helix transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
angiogenesis
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
locomotory behavior
spinal cord association neuron differentiation
hemopoiesis
erythrocyte differentiation
megakaryocyte differentiation
platelet formation
basophil differentiation
positive regulation of protein complex assembly
embryonic hemopoiesis
megakaryocyte development
regulation of cell proliferation
erythrocyte maturation
cell fate commitment
positive regulation of erythrocyte differentiation
positive regulation of chromatin assembly or disassembly
positive regulation of transcription, DNA-templated
positive regulation of mitotic cell cycle
positive regulation of transcription from RNA polymerase II promoter
positive regulation of cell division
astrocyte fate commitment
definitive hemopoiesis
hemangioblast cell differentiation
hematopoietic stem cell differentiation
regulation of mast cell differentiation
regulation of stem cell population maintenance
RNA polymerase II regulatory region sequence-specific DNA binding
RNA polymerase II core promoter sequence-specific DNA binding
RNA polymerase II distal enhancer sequence-specific DNA binding
RNA polymerase II transcription factor activity, sequence-specific DNA binding
RNA polymerase II transcription factor binding
chromatin binding
transcription factor activity, sequence-specific DNA binding
protein binding
enzyme binding
histone deacetylase binding
transcription regulatory region DNA binding
protein heterodimerization activity
E-box binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0