This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06629738_g1
See other ATP5O GE Assays ›
Gene Symbol:
ATP5O
Gene Name:
ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit
Gene Aliases:
ATPO, HMC08D05, OSCP
Chromosome Location:
Chr.21: 33903453 - 33915854 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06629738_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5O
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001697.2 NP_001688.1 5 467 67
GenBank mRNA AF088071.1 - 3 262 67
AK124187.1 - 4 1608 67
AK222608.1 - 5 399 67
AK222962.1 - 5 394 67
AK311796.1 - 5 394 67
AV734214.1 - 5 463 67
BC021233.2 - 5 380 4126036 67
BC022865.1 - 5 398 4103001 67
BT019836.1 - 5 376 67
CD174837.1 - 5 437 67
CR456822.1 - 5 376 67
X83218.1 - 5 412 67
X83219.1 - 2 178 67
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DQ656027.1 - 3 Chr. 21 - 33584687 - 33912331 [+] on Build GRCh38 67

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATP5O

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
proton transport
mitochondrial ATP synthesis coupled proton transport
transporter activity
protein binding
drug binding
ATPase activity
transmembrane transporter activity
proton-transporting ATP synthase activity, rotational mechanism

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0