This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06629851_s1
See other COMT GE Assays ›
Gene Symbol:
COMT ARVCF
Gene Name:
catechol-O-methyltransferase
armadillo repeat gene deleted in velocardiofacial syndrome
Gene Aliases:
HEL-S-98n
-
Chromosome Location:
Chr.22: 19941740 - 19969975 on Build GRCh38
Chromosome Location:
Chr.22: 19966491 - 20016802 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
-
Assay ID Hs06629851_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: COMT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000754.3 NP_000745.1 6 1153 56
NM_001135161.1 NP_001128633.1 6 1111 56
NM_001135162.1 NP_001128634.1 6 1128 56
NM_007310.2 NP_009294.1 4 884 56
XM_011529886.1 XP_011528188.1 6 1058 56
XM_017028594.1 XP_016884083.1 6 1377 56
XM_017028595.1 XP_016884084.1 6 1321 56
GenBank mRNA AK130031.1 - 7 934 56
AK290440.1 - 6 1092 56
BC000419.2 - 6 1082 2964400 56
BC005867.2 - 6 1082 2964400 56
BC011935.2 - 5 997 3503220 56
BC100018.1 - 6 1143 6163659 56
EF212883.1 - 3 308 56
EF217321.1 - 2 228 56
EF217322.1 - 2 255 56
FJ224345.1 - 6 1082 56
M58525.1 - 6 1109 56
M65212.1 - 5 982 56
M65213.1 - 1 154 56
Gene Symbol: ARVCF
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AL390148.1 - 1 56

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene COMT
Click here to find siRNAs for Gene ARVCF

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

female pregnancy
learning
short-term memory
estrogen metabolic process
response to organic cyclic compound
cellular response to phosphate starvation
methylation
response to lipopolysaccharide
developmental process
negative regulation of renal sodium excretion
neurotransmitter catabolic process
dopamine catabolic process
response to drug
negative regulation of dopamine metabolic process
response to pain
multicellular organismal reproductive process
negative regulation of smooth muscle cell proliferation
positive regulation of homocysteine metabolic process
regulation of sensory perception of pain
magnesium ion binding
protein binding
methyltransferase activity
O-methyltransferase activity
catechol O-methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0