This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06629892_s1
See other HMOX1 GE Assays ›
Gene Symbol:
HMOX1
Gene Name:
heme oxygenase 1
Gene Aliases:
HMOX1D, HO-1, HSP32, bK286B10
Chromosome Location:
Chr.22: 35381067 - 35394214 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06629892_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HMOX1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002133.2 NP_002124.1 3 305 103
GenBank mRNA AF051791.1 - 2 177 103
AK313120.1 - 3 275 103
BC001491.2 - 3 264 3504480 103
BP288767.1 - 3 305 103
BT019785.1 - 2 198 103
CR456505.1 - 3 313 103
CR541945.1 - 3 198 103
CR541968.1 - 3 198 103
JF323038.1 - 3 202 103
M23041.1 - 3 230 103
X06985.1 - 3 278 103

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HMOX1

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

angiogenesis
endothelial cell proliferation
wound healing involved in inflammatory response
negative regulation of leukocyte migration
heme oxidation
cellular iron ion homeostasis
response to oxidative stress
small GTPase mediated signal transduction
excretion
regulation of blood pressure
cell death
intrinsic apoptotic signaling pathway in response to DNA damage
negative regulation of muscle cell apoptotic process
smooth muscle hyperplasia
cellular response to nutrient
negative regulation of mast cell cytokine production
erythrocyte homeostasis
low-density lipoprotein particle clearance
regulation of transcription from RNA polymerase II promoter in response to iron
cellular response to heat
response to nicotine
intracellular signal transduction
heme catabolic process
response to hydrogen peroxide
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of mast cell degranulation
negative regulation of DNA binding
negative regulation of sequence-specific DNA binding transcription factor activity
negative regulation of neuron apoptotic process
regulation of transcription from RNA polymerase II promoter in response to oxidative stress
response to estrogen
positive regulation of chemokine biosynthetic process
regulation of angiogenesis
positive regulation of angiogenesis
positive regulation of vasodilation
positive regulation of smooth muscle cell proliferation
negative regulation of smooth muscle cell proliferation
regulation of sequence-specific DNA binding transcription factor activity
protein homooligomerization
iron ion homeostasis
cellular response to arsenic-containing substance
cellular response to cadmium ion
cellular response to hypoxia
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
heme oxygenase (decyclizing) activity
phospholipase D activity
signal transducer activity
protein binding
enzyme binding
heme binding
protein homodimerization activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0