This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06630946_g1
See other ATP5G3 GE Assays ›
Gene Symbol:
ATP5G3
Gene Name:
ATP synthase, H+ transporting, mitochondrial Fo complex subunit C3 (subunit 9)
Gene Aliases:
P3
Chromosome Location:
Chr.2: 175176258 - 175181762 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06630946_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5G3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001002258.4 NP_001002258.1 3 508 96
NM_001190329.1 NP_001177258.1 4 418 96
NM_001689.4 NP_001680.1 4 418 96
GenBank mRNA AK125917.1 - 4 3263 96
AK311999.1 - 4 351 96
BC106881.2 - 3 333 40029441 96
BF210704.1 - 4 420 96
BI756002.1 - 4 381 96
BI765737.1 - 4 387 96
BQ054164.1 - 4 377 96
CA424417.1 - 2 96
CR407601.1 - 3 238 96
U09813.1 - 3 492 96

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene ATP5G3

Panther Classification:

Molecular Function -

ATP synthase

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
aging
ATP hydrolysis coupled proton transport
mitochondrial ATP synthesis coupled proton transport
response to ethanol
transporter activity
lipid binding
hydrogen ion transmembrane transporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0