This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06631724_s1
See other OGG1 GE Assays ›
Gene Symbol:
OGG1
Gene Name:
8-oxoguanine DNA glycosylase
Gene Aliases:
HMMH, HOGG1, MUTM, OGH1
Chromosome Location:
Chr.3: 9749944 - 9788246 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06631724_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OGG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002542.5 NP_002533.1 5 1166 59
NM_016819.3 NP_058212.1 5 1166 59
NM_016820.3 NP_058213.1 5 1166 59
NM_016821.2 NP_058214.1 5 1166 59
NM_016828.2 NP_058437.1 5 1166 59
NM_016829.2 NP_058438.1 5 1166 59
XM_011533760.1 XP_011532062.1 5 1161 59
XM_017006496.1 XP_016861985.1 5 1161 59
GenBank mRNA AB000410.1 - 5 1091 59
AB019530.1 - 4 582 59
AB019531.1 - 4 582 59
AB019532.1 - 3 422 59
AB037878.1 - 2 123 59
AB037882.1 - 2 123 59
AF003595.1 - 5 879 59
AF026691.1 - 5 1030 59
AK289858.1 - 5 1119 59
BC000657.2 - 5 1120 3350168 59
U88527.1 - 5 1133 59
U88620.1 - 5 1133 59
U96710.1 - 5 880 59
Y11731.2 - 5 1131 59
Y11838.1 - 5 1130 59
Y13277.1 - 5 1127 59

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene OGG1

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

acute inflammatory response
base-excision repair
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
response to oxidative stress
aging
response to radiation
response to light stimulus
response to estradiol
regulation of protein import into nucleus, translocation
nucleotide-excision repair, DNA incision
response to drug
negative regulation of apoptotic process
depurination
depyrimidination
response to ethanol
response to folic acid
cellular response to cadmium ion
negative regulation of double-strand break repair via single-strand annealing
damaged DNA binding
endonuclease activity
protein binding
microtubule binding
oxidized purine nucleobase lesion DNA N-glycosylase activity
oxidized purine DNA binding
8-oxo-7,8-dihydroguanine DNA N-glycosylase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline