This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs06631737_s1
See other OGG1 GE Assays ›
Gene Symbol:
OGG1
Gene Name:
8-oxoguanine DNA glycosylase
Gene Aliases:
HMMH, HOGG1, MUTM, OGH1
Chromosome Location:
Chr.3: 9749944 - 9788246 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs06631737_s1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OGG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_002542.5 NP_002533.1 7 1349 53
NM_016819.3 NP_058212.1 6 1593 53
NM_016820.3 NP_058213.1 7 1366 53
XM_017006493.1 XP_016861982.1 6 1193 53
XM_017006495.1 XP_016861984.1 6 1210 53
GenBank mRNA AB000410.1 - 7 1274 53
AB019532.1 - 4 849 53
AB037878.1 - 4 306 53
AB037879.1 - 4 337 53
AB037880.1 - 3 155 53
AB037881.1 - 2 399 53
AB037882.1 - 4 323 53
AF003595.1 - 7 1062 53
AF026691.1 - 7 1231 53
AK289858.1 - 7 1302 53
BC000657.2 - 7 1303 3350168 53
U88527.1 - 7 1316 53
U96710.1 - 7 1081 53
Y11731.2 - 7 1314 53
Y11838.1 - 7 1313 53

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene OGG1

Panther Classification:

Molecular Function -

DNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

acute inflammatory response
base-excision repair
regulation of transcription, DNA-templated
cellular response to DNA damage stimulus
response to oxidative stress
aging
response to radiation
response to light stimulus
response to estradiol
regulation of protein import into nucleus, translocation
nucleotide-excision repair, DNA incision
response to drug
negative regulation of apoptotic process
depurination
depyrimidination
response to ethanol
response to folic acid
cellular response to cadmium ion
negative regulation of double-strand break repair via single-strand annealing
damaged DNA binding
endonuclease activity
protein binding
microtubule binding
oxidized purine nucleobase lesion DNA N-glycosylase activity
oxidized purine DNA binding
8-oxo-7,8-dihydroguanine DNA N-glycosylase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0